This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06017864_g1
See other PNPLA4 GE Assays ›
Gene Symbol:
PNPLA4
Gene Name:
patatin like phospholipase domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.X: 5725448 - 5755286 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06017864_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNPLA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002762600.4 XP_002762646.1 3 - 4 477 106
XM_008988911.2 XP_008987159.1 3 - 4 326 106
XM_008988914.2 XP_008987162.1 2 - 3 1424 106
XM_008988916.2 XP_008987164.1 2 - 3 144 106
XM_017968619.1 XP_017824108.1 2 - 3 1435 106
XM_017968620.1 XP_017824109.1 3 - 4 496 106
XM_017968621.1 XP_017824110.1 3 - 4 392 106
XM_017968622.1 XP_017824111.1 3 - 4 326 106
XM_017968623.1 XP_017824112.1 3 - 4 1252 106
XM_017968625.1 XP_017824114.1 2 - 3 1438 106
XM_017968626.1 XP_017824115.1 2 - 3 224 106
XM_017968627.1 XP_017824116.1 2 - 3 144 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008988916.2, XM_017968626.1, XM_017968627.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fn2v6:80/100.66.76.37:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0