Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979120.2 | XP_008977368.1 | 9 | 3043 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10D| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979120.2 | XP_008977368.1 | 9 | 3043 | 85 | |
Set Membership: |
|