Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755957.4 | XP_002756003.1 | 6 - 7 | 1729 | 85 | |
| XM_009009265.2 | XP_009007513.1 | 7 - 8 | 1637 | 85 | ||
| XM_009009267.2 | XP_009007515.1 | 5 - 6 | 1585 | 85 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map