This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06058001_m1
See other ACMSD GE Assays ›
Gene Symbol:
ACMSD
Gene Name:
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.6: 85087304 - 85177902 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06058001_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACMSD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002749492.2 XP_002749538.1 5 - 6 620 62
XM_008998984.2 XP_008997232.1 5 - 6 602 62
XM_008998985.2 XP_008997233.1 5 - 6 603 62
XM_008998986.2 XP_008997234.1 5 - 6 603 62
XM_008998987.2 XP_008997235.1 4 - 5 510 62
XM_017973515.1 XP_017829004.1 4 - 5 557 62
XM_017973516.1 XP_017829005.1 5 - 6 605 62
XM_017973517.1 XP_017829006.1 5 - 6 603 62
XM_017973518.1 XP_017829007.1 3 - 4 408 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

decarboxylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0