This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06075180_m1
See other GPRC6A GE Assays ›
Gene Symbol:
GPRC6A
Gene Name:
G protein-coupled receptor class C group 6 member A
Gene Aliases:
-
Chromosome Location:
Chr.4: 116664423 - 116699329 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06075180_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002746896.2 XP_002746942.1 1 - 2 229 71
XM_002746897.2 XP_002746943.1 1 - 2 229 71
XM_002746898.2 XP_002746944.1 1 - 2 229 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-2vf2v:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline