This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06094000_m1
See other ETFBKMT GE Assays ›
Gene Symbol:
ETFBKMT
Gene Name:
electron transfer flavoprotein subunit beta lysine methyltransferase
Gene Aliases:
METTL20
Chromosome Location:
Chr.9: 35745506 - 35766827 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06094000_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETFBKMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009003669.2 XP_009001917.1 3 - 4 742 88
XM_009003670.2 XP_009001918.1 3 - 4 630 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0