This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06098225_g1
See other MILR1 GE Assays ›
Gene Symbol:
MILR1
Gene Name:
mast cell immunoglobulin like receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 83193815 - 83254276 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06098225_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MILR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008996919.2 XP_008995167.1 7 - 8 1044 88
XM_008996920.2 XP_008995168.1 7 - 8 1044 88
XM_008996923.2 XP_008995171.1 6 - 7 954 88
XM_017972469.1 XP_017827958.1 7 - 8 1051 88
XM_017972470.1 XP_017827959.1 6 - 7 786 88
XM_017972471.1 XP_017827960.1 6 - 7 767 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0