This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06123498_g1
See other MUTYH GE Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.7: 80150061 - 80163107 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06123498_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MUTYH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009001122.1 XP_008999370.1 4 - 5 456 100
XM_009001123.1 XP_008999371.1 4 - 5 462 100
XM_009001125.2 XP_008999373.1 4 - 5 406 100
XM_017974663.1 XP_017830152.1 4 - 5 406 100
XM_017974664.1 XP_017830153.1 4 - 5 406 100
XM_017974665.1 XP_017830154.1 4 - 5 406 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline