This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06131488_s1
See other PPIL3 GE Assays ›
Gene Symbol:
PPIL3
Gene Name:
peptidylprolyl isomerase like 3
Gene Aliases:
-
Chromosome Location:
Chr.6: 116384320 - 116405512 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06131488_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008999182.2 XP_008997430.1 7 1120 105
XM_017973619.1 XP_017829108.1 6 865 105
XM_017973620.1 XP_017829109.1 7 945 105
XM_017973621.1 XP_017829110.1 7 934 105
XM_017973622.1 XP_017829111.1 7 1219 105
XM_017973623.1 XP_017829112.1 7 958 105
XM_017973624.1 XP_017829113.1 7 863 105
XM_017973625.1 XP_017829114.1 7 863 105
XM_017973626.1 XP_017829115.1 7 863 105
XM_017973627.1 XP_017829116.1 7 865 105
XM_017973628.1 XP_017829117.1 7 863 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline