This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cp03756137_s1
See other HPGD GE Assays ›
Gene Symbol:
Hpgd
Gene Name:
15-hydroxyprostaglandin dehydrogenase
Gene Aliases:
PGDH, PGDH1
Chromosome Location:
Chr.Un|NT_176383: 18145078 - 18180289 on Build Cavpor3.0
Species:
Guinea pig
Species Specific ID (Flybase ID):
-
Assay ID Cp03756137_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hpgd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172961.1 NP_001166432.1 5 454 78
GenBank mRNA Y07953.1 - 5 454 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Cavia porcellus Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bm6jx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0