This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cp04183084_m1
See other RARB GE Assays ›
Gene Symbol:
Rarb
Gene Name:
retinoic acid receptor beta
Gene Aliases:
nRARs
Chromosome Location:
Chr.Un|NT_176419: 71056380 - 71217713 on Build Cavpor3.0
Species:
Guinea pig
Species Specific ID (Flybase ID):
-
Assay ID Cp04183084_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rarb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172731.1 NP_001166202.1 4 - 5 1028 60
XM_005008210.2 XP_005008267.1 4 - 5 903 60
XM_013143710.2 XP_012999164.1 4 - 5 628 60
GenBank mRNA AY280717.2 - 4 - 5 1028 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Cavia porcellus Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0