Assay Details
| Gene Symbol: mim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201940.3 | NP_001188869.1 | 15 - 16 | 6864 | 70 | |
| NM_001259230.2 | NP_001246159.1 | 16 - 17 | 6384 | 70 | ||
| NM_001273819.1 | NP_001260748.1 | 17 - 18 | 6186 | 70 | ||
More Information
Set Membership: |
|

mim
Genomic Map