This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01797997_m1
See other GOT1 GE Assays ›
Gene Symbol:
Got1
Gene Name:
Glutamate oxaloacetate transaminase 1
Gene Aliases:
BEST:GH07823, CG8430, Dmel\CG8430, Dmel_CG8430, FBgn 1124, GOT-1, Got-1, Got2, got1
Chromosome Location:
Chr.2R: 16138352 - 16141928 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0001124
Assay ID Dm01797997_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Got1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_137242.3 NP_611086.1 1 - 2 206 63
NM_166148.2 NP_725534.1 2 - 3 205 63
GenBank mRNA AY061313.1 - 1 - 2 179 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

aspartate metabolic process
aspartate biosynthetic process
glutamate biosynthetic process
L-aspartate:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0