This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01798120_m1
See other ATOX1 GE Assays ›
Gene Symbol:
Atox1
Gene Name:
Antioxidant 1 copper chaperone
Gene Aliases:
CG12562, CG32446, DmAtox1, Dmel\CG32446, Dmel_CG32446
Chromosome Location:
Chr.3L: 21640995 - 21642599 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0052446
Assay ID Dm01798120_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275255.1 NP_001262184.1 2 - 3 272 89
NM_168932.2 NP_730672.1 2 - 3 272 89
GenBank mRNA AY061200.1 - 2 - 3 222 89
KX532007.1 - 1 - 2 77 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

cellular copper ion homeostasis
response to oxidative stress
metal ion transport
copper ion homeostasis
copper chaperone activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0