This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01798159_m1
See other CSP GE Assays ›
Gene Symbol:
Csp
Gene Name:
Cysteine string protein
Gene Aliases:
CG6395, CSP, CSP32, Cys, D-CSP, D.M.CSP, DCSP-1, DNAJC5, DSP/DNAJC5, Dcsp, Dmel\CG6395, Dmel_CG6395, MAB ab49, ab49, anon-WO02059370.50, csp, csp29, csp32, l(3)03988
Chromosome Location:
Chr.3L: 22267382 - 22273745 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004179
Assay ID Dm01798159_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Csp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275277.1 NP_001262206.1 4 - 5 640 61
NM_001275278.1 NP_001262207.1 4 - 5 640 61
NM_001300215.1 NP_001287144.1 4 - 5 826 61
NM_168949.2 NP_730713.1 4 - 5 640 61
GenBank mRNA BT100236.1 - 4 - 5 618 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s)

protein folding
exocytosis
synaptic vesicle exocytosis
neuron cellular homeostasis
regulation of neuromuscular synaptic transmission
negative regulation of response to wounding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0