This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01800875_g1
See other EOGT GE Assays ›
Gene Symbol:
Eogt
Gene Name:
EGF-domain O-GlcNAc transferase
Gene Aliases:
CG9867, Dmel\CG9867, Dmel_CG9867, eogt
Chromosome Location:
Chr.2L: 2381097 - 2383073 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264672
Assay ID Dm01800875_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eogt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273005.1 NP_001259934.1 2 - 3 1139 77
NM_134834.3 NP_608678.1 1 - 2 1202 77
GenBank mRNA AB675601.1 - 1 - 2 965 77
AY058292.1 - 1 - 2 1159 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein O-linked glycosylation
larval chitin-based cuticle development
protein O-GlcNAcylation via threonine
protein N-acetylglucosaminyltransferase activity
transferase activity, transferring glycosyl groups
protein O-GlcNAc transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0