This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01801488_g1
See other ELP3 GE Assays ›
Gene Symbol:
Elp3
Gene Name:
Elongator complex protein 3
Gene Aliases:
CG15433, D-elp3, Dmel/ELP3, Dmel\CG15433, Dmel\ELP3, Dmel_CG15433, ELP3, HAT, dmHAG408, elp-3, elp3
Chromosome Location:
Chr.2L: 4444732 - 4446765 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031604
Assay ID Dm01801488_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134990.3 NP_608834.1 3 - 4 1213 70
GenBank mRNA AY089621.1 - 3 - 4 1211 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

instar larval development
tRNA wobble base 5-methoxycarbonylmethyl-2-thiouridinylation
regulation of transcription from RNA polymerase II promoter
locomotory behavior
histone acetylation
peptidyl-lysine acetylation
hemopoiesis
sleep
cytoskeletal matrix organization at active zone
synapse organization
synaptic growth at neuromuscular junction
regulation of photoreceptor cell axon guidance
phosphorylase kinase regulator activity
acetyltransferase activity
iron-sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0