This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01805782_g1
See other AGPAT3 GE Assays ›
Gene Symbol:
Agpat3
Gene Name:
1-Acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
AGPAT, AGPAT3, CG4729, Dmel\CG4729, Dmel_CG4729
Chromosome Location:
Chr.3L: 16381772 - 16385957 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0036623
Assay ID Dm01805782_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259864.1 NP_001246793.1 4 - 5 1460 88
NM_140631.2 NP_648888.1 4 - 5 1216 88
NM_168661.2 NP_730160.1 4 - 5 1283 88
NM_168662.2 NP_730161.1 4 - 5 1287 88
NM_168663.2 NP_730162.1 2 - 3 456 88
GenBank mRNA AY051615.1 - 2 - 3 456 88
BT001546.1 - 4 - 5 1216 88
BT124744.1 - 4 - 5 1460 88
KX531900.1 - 1 - 2 343 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Process(es)

1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0