This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01806606_g1
See other NAXD GE Assays ›
Gene Symbol:
Naxd
Gene Name:
NAD(P)HX dehydratase
Gene Aliases:
CG10424, Dmel\CG10424, Dmel_CG10424
Chromosome Location:
Chr.3L: 19096399 - 19097709 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0036848
Assay ID Dm01806606_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naxd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_140833.3 NP_649090.1 1 - 2 398 91
GenBank mRNA AY060665.1 - 1 - 2 359 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

dehydratase lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

metabolite repair
ATP-dependent NAD(P)H-hydrate dehydratase activity
ADP-dependent NAD(P)H-hydrate dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0