This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01808344_g1
See other AASDH GE Assays ›
Gene Symbol:
Aasdh
Gene Name:
Aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
CG13401, Dmel\CG13401, Dmel_CG13401, U26
Chromosome Location:
Chr.2L: 8445861 - 8449576 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0027780
Assay ID Dm01808344_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aasdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001298825.1 NP_001285754.1 2 - 3 755 58
NM_135386.5 NP_609230.2 2 - 3 755 58
NM_205938.2 NP_995660.2 2 - 3 755 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
catalytic activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0