This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809484_m1
See other WWOX GE Assays ›
Gene Symbol:
Wwox
Gene Name:
WW domain containing oxidoreductase
Gene Aliases:
CG7221, DmWWOX, Dmel\CG7221, Dmel_CG7221, WWOX, anon-EST:Posey177
Chromosome Location:
Chr.2L: 7999670 - 8004286 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031972
Assay ID Dm01809484_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wwox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001298806.1 NP_001285735.1 3 - 4 879 60
NM_135327.4 NP_609171.1 3 - 4 879 60
GenBank mRNA AY119574.1 - 3 - 4 819 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s)

response to ionizing radiation
defense response to Gram-negative bacterium
regulation of reactive oxygen species metabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0