This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01810764_mH
See other PDE1C GE Assays ›
Gene Symbol:
Pde1c
Gene Name:
Phosphodiesterase 1c
Gene Aliases:
CG14940, CG14942, CG14943, CG14944, CG31757, CG31758, CG42325, CG44007, Dmel\CG44007, Dmel_CG44007, PDE1, PDE1C, PDE1c, mdcds_1440
Chromosome Location:
Chr.2L: 11814156 - 11928572 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264815
Assay ID Dm01810764_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144344.2 NP_001137816.1 9 - 10 3142 67
NM_001144345.3 NP_001137817.1 8 - 9 3788 67
NM_001144346.3 NP_001137818.1 7 - 8 3483 67
NM_001144347.3 NP_001137819.2 8 - 9 3296 67
NM_001144348.4 NP_001137820.1 9 - 10 3155 67
NM_001259065.2 NP_001245994.1 6 - 7 2928 67
NM_001259066.2 NP_001245995.2 8 - 9 3119 67
NM_001273466.1 NP_001260395.1 8 - 9 3546 67
NM_135676.6 NP_609520.1 3 - 4 2012 67
GenBank mRNA AY118396.1 - 3 - 4 2011 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
multicellular organism reproduction
cAMP metabolic process
cGMP metabolic process
male mating behavior
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin binding
3',5'-cyclic-GMP phosphodiesterase activity
calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline