This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01813257_g1
See other FH GE Assays ›
Gene Symbol:
fh
Gene Name:
frataxin
Gene Aliases:
BcDNA:AT09528, CG8971, DFH, Dfh, Dmel\CG8971, Dmel_CG8971, X25, dfh
Chromosome Location:
Chr.X: 9148080 - 9149071 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0030092
Assay ID Dm01813257_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_078539.3 NP_511094.1 1 - 2 377 55
GenBank mRNA AJ002208.1 - 1 - 2 340 55
AY094649.1 - 1 - 2 365 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

cellular iron ion homeostasis
response to oxidative stress
mitochondrion organization
determination of adult lifespan
adult locomotory behavior
response to iron ion
lipid catabolic process
iron-sulfur cluster assembly
iron incorporation into metallo-sulfur cluster
axonal transport of mitochondrion
response to hydrogen peroxide
positive regulation of heart contraction
positive regulation of ecdysteroid biosynthetic process
regulation of mitochondrial membrane potential
oxidation-reduction process
positive regulation of oxidative phosphorylation
ferroxidase activity
ferrous iron binding
ferric iron binding
iron chaperone activity
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0