This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01817650_g1
See other RPL13 GE Assays ›
Gene Symbol:
RpL13
Gene Name:
Ribosomal protein L13
Gene Aliases:
Bbc1, CG4651, Dmbbc1, Dmel\CG4651, Dmel_CG4651, L13, Rp L13, Rpl13, anon-EST:fe1D1, bbc1, rpL13
Chromosome Location:
Chr.2L: 9967313 - 9968645 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0011272
Assay ID Dm01817650_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RpL13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038802.2 NP_001033891.1 2 - 3 290 58
NM_001273383.2 NP_001260312.1 1 - 2 548 58
NM_078806.4 NP_523530.1 2 - 3 307 58
GenBank mRNA BT053725.1 - 1 - 2 261 58
BT128812.1 - 2 - 3 292 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translation
translation
RNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline