This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01818967_g1
See other GSTT1 GE Assays ›
Gene Symbol:
GstT1
Gene Name:
Glutathione S transferase T1
Gene Aliases:
BcDNA:RE63575, CG12930, CG12930B, CG30000, DmGSTT1, Dmel\CG30000, Dmel_CG30000, GSTT1, gstt1
Chromosome Location:
Chr.2R: 9605988 - 9607184 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0050000
Assay ID Dm01818967_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GstT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_136665.5 NP_610509.2 1 - 2 216 95
GenBank mRNA AY070580.1 - 1 - 2 214 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
glutathione transferase activity
glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-qnxks:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0