This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01819036_g1
See other VAMP7 GE Assays ›
Gene Symbol:
Vamp7
Gene Name:
Vesicle-associated membrane protein 7
Gene Aliases:
CG1599, Dmel\CG1599, Dmel_CG1599, TI-VAMP, VAMP7, VAMP7/8, dVamp7, vamp7
Chromosome Location:
Chr.2R: 9841080 - 9842502 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0266186
Assay ID Dm01819036_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vamp7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299326.1 NP_001286255.1 2 - 3 580 109
NM_136680.4 NP_610524.1 2 - 3 522 109
GenBank mRNA AY071695.1 - 2 - 3 534 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
vesicle fusion
endosome to lysosome transport
autophagosome maturation
SNARE binding
SNAP receptor activity
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0