This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01819966_g1
See other DERA GE Assays ›
Gene Symbol:
Dera
Gene Name:
Deoxyribose-phosphate aldolase
Gene Aliases:
CG8525, Dmel\CG8525, Dmel_CG8525
Chromosome Location:
Chr.2R: 12425483 - 12426975 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033735
Assay ID Dm01819966_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dera
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299419.1 NP_001286348.1 2 - 3 755 57
NM_136937.3 NP_610781.1 2 - 3 864 57
GenBank mRNA AY061335.1 - 2 - 3 698 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

deoxyribonucleotide catabolic process
carbohydrate catabolic process
deoxyribose-phosphate aldolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline