This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01822042_m1
See other GAD1 GE Assays ›
Gene Symbol:
Gad1
Gene Name:
Glutamic acid decarboxylase 1
Gene Aliases:
CG14994, DGAD1, DGad1, Dmel\CG14994, Dmel_CG14994, GAD, GAD-1, GAD1, GAD[[1]], Gad, Glb, dGad1, dgad1, gad, gad1, l(3)64An, l(3)SH10
Chromosome Location:
Chr.3L: 4074077 - 4088744 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004516
Assay ID Dm01822042_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gad1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001274467.1 NP_001261396.1 2 - 3 252 59
NM_079190.3 NP_523914.2 2 - 3 285 59
NM_168055.2 NP_728930.1 2 - 3 597 59
NM_168056.2 NP_728931.1 3 - 4 386 59
GenBank mRNA AY089526.1 - 2 - 3 305 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

glutamate catabolic process
synapse assembly
neuromuscular junction development
larval locomotory behavior
olfactory learning
gamma-aminobutyric acid biosynthetic process
response to mechanical stimulus
neurotransmitter receptor metabolic process
glutamate decarboxylase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0