This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01822506_g1
See other RPS9 GE Assays ›
Gene Symbol:
RpS9
Gene Name:
Ribosomal protein S9
Gene Aliases:
CG3395, Dmel\CG3395, Dmel_CG3395, RPS9, Rp S9, RpL11, S9, chr3L:9483841..9483937, chr3L:9484246..9484474
Chromosome Location:
Chr.3L: 9509793 - 9511531 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0010408
Assay ID Dm01822506_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RpS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300079.1 NP_001287008.1 1 - 2 247 84
NM_079280.4 NP_524004.2 2 - 3 275 84
NM_168350.2 NP_729506.1 1 - 2 247 84
NM_168351.2 NP_729507.1 2 - 3 275 84
GenBank mRNA AY061396.1 - 1 - 2 250 84
BT088955.1 - 1 - 2 249 84
KX531268.1 - 1 - 2 220 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translation
positive regulation of translational fidelity
structural constituent of ribosome
rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline