This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01823209_g1
See other IDE GE Assays ›
Gene Symbol:
Ide
Gene Name:
Insulin degrading metalloproteinase
Gene Aliases:
CG5517, Dmel\CG5517, Dmel_CG5517, IDE, dIDE, dIde, dide, dlde, ide
Chromosome Location:
Chr.3L: 20371672 - 20375896 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0001247
Assay ID Dm01823209_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ide
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079458.4 NP_524182.3 5 - 6 1052 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
determination of adult lifespan
peptide catabolic process
positive regulation of innate immune response
negative regulation of growth
negative regulation of insulin receptor signaling pathway
regulation of oviposition
negative regulation of developmental growth
defense response to Gram-negative bacterium
proteolysis involved in cellular protein catabolic process
regulation of trehalose metabolic process
insulin catabolic process
metalloendopeptidase activity
metallopeptidase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline