This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01823896_g1
See other EIF3M GE Assays ›
Gene Symbol:
eIF3m
Gene Name:
eukaryotic translation initiation factor 3 subunit m
Gene Aliases:
CG8309, Dmel\CG8309, Dmel_CG8309, TANGO7, Tango 7, Tango7, Tng7, anon-EST:Posey67, anon-EST:Posey76, eIF3M, tango7
Chromosome Location:
Chr.2R: 14163066 - 14164825 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033902
Assay ID Dm01823896_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF3m
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299478.1 NP_001286407.1 2 - 3 311 78
NM_137088.4 NP_610932.1 2 - 3 515 78
GenBank mRNA AY118924.1 - 2 - 3 299 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational initiation
translation
Golgi organization
sperm individualization
protein secretion
activation of cysteine-type endopeptidase activity
positive regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
translation initiation factor activity
translation initiation factor binding
caspase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0