This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01828038_g1
See other HDAC4 GE Assays ›
Gene Symbol:
HDAC4
Gene Name:
Histone deacetylase 4
Gene Aliases:
CG1770, DHDAC4, Dmel\CG1770, Dmel_CG1770, GC1770, HDAC, HDAC4a, dHDAC4, dmHDA405, hdac4
Chromosome Location:
Chr.X: 13262687 - 13285632 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0041210
Assay ID Dm01828038_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014736.5 NP_001014736.2 5 - 6 965 57
NM_001258722.3 NP_001245651.1 6 - 7 1006 57
NM_001272578.2 NP_001259507.1 5 - 6 1327 57
NM_001272579.2 NP_001259508.1 4 - 5 1163 57
NM_001272580.2 NP_001259509.1 4 - 5 1212 57
NM_001272581.2 NP_001259510.1 4 - 5 901 57
NM_132640.6 NP_572868.3 5 - 6 965 57
NM_167356.3 NP_727682.1 3 - 4 701 57
GenBank mRNA BT133402.1 - 5 - 6 654 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
memory
long-term memory
insulin receptor signaling pathway
cellular response to starvation
histone acetylation
histone deacetylation
positive regulation of sequence-specific DNA binding transcription factor activity
histone deacetylase activity
SUMO transferase activity
protein deacetylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0