This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01830335_g1
See other DNR1 GE Assays ›
Gene Symbol:
dnr1
Gene Name:
defense repressor 1
Gene Aliases:
CG12489, DNR1, Dmel\CG12489, Dmel_CG12489, Dnr-1, Dnr1, dNR1
Chromosome Location:
Chr.2R: 22563152 - 22592964 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0260866
Assay ID Dm01830335_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001274208.2 NP_001261137.1 3 - 4 2280 115
NM_137836.3 NP_611680.2 3 - 4 2280 115
GenBank mRNA AY095032.1 - 3 - 4 2268 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of biosynthetic process of antibacterial peptides active against Gram-negative bacteria
protein ubiquitination
sleep
ubiquitin-protein transferase activity
zinc ion binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0