This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01830529_s1
See other CNBP GE Assays ›
Gene Symbol:
CNBP
Gene Name:
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases:
BEST:GM06241, CG11716, CG3800, Dmel\CG3800, Dmel_CG3800, anon-EST:fe1B4, anon-EST:fe2B6, dCNBP
Chromosome Location:
Chr.2R: 22934569 - 22937077 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0034802
Assay ID Dm01830529_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001202063.2 NP_001188992.1 2 1970 115
NM_137895.3 NP_611739.1 1 2411 115
GenBank mRNA AY118596.1 - 1 2011 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytoplasmic translation
single-stranded RNA binding
mRNA binding
zinc ion binding
translation regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0