Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 6 - 7 | 2358 | 70 | |
| NM_001104057.1 | NP_001097527.1 | 7 - 8 | 2577 | 70 | ||
| GenBank mRNA | AB061681.1 | - | 6 - 7 | 2357 | 70 | |
More Information
Set Membership: |
|

form3
Genomic Map