This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01833360_g1
See other C3G GE Assays ›
Gene Symbol:
C3G
Gene Name:
C3G guanyl-nucleotide exchange factor
Gene Aliases:
C(3)G, CG14436, CG3126, CG42328, DC3-G, DC3G, Dmel\CG42328, Dmel_CG42328, c(3)G
Chromosome Location:
Chr.X: 6780790 - 6803593 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0259228
Assay ID Dm01833360_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C3G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144694.2 NP_001138166.1 14 - 15 4328 116
NM_001144695.3 NP_001138167.1 12 - 13 4582 116
NM_001144696.3 NP_001138168.1 13 - 14 4735 116
NM_001144697.2 NP_001138169.1 13 - 14 4175 116
NM_001272362.1 NP_001259291.1 13 - 14 4579 116
NM_001272363.1 NP_001259292.1 12 - 13 3958 116
NM_001272364.1 NP_001259293.1 13 - 14 4735 116
NM_001298037.1 NP_001284966.1 12 - 13 4408 116
NM_132122.5 NP_572350.2 13 - 14 5398 116
NM_176694.3 NP_788867.1 13 - 14 5858 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
somatic muscle development
muscle attachment
regulation of GTPase activity
positive regulation of GTPase activity
sarcomere organization
positive regulation of smoothened signaling pathway
Ras guanyl-nucleotide exchange factor activity
Rap guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0