This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01834505_m1
See other GALE GE Assays ›
Gene Symbol:
Gale
Gene Name:
UDP-galactose 4'-epimerase
Gene Aliases:
CG12030, Dmel\CG12030, Dmel_CG12030, GALE, dGALE
Chromosome Location:
Chr.3L: 648555 - 651311 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0035147
Assay ID Dm01834505_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gale
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259608.2 NP_001246537.1 2 - 3 519 51
NM_138200.4 NP_612044.1 2 - 3 738 51
GenBank mRNA AY058582.1 - 2 - 3 764 51

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

galactose metabolic process
galactose catabolic process via UDP-galactose
larval lymph gland hemopoiesis
UDP-N-acetylglucosamine 4-epimerase activity
UDP-glucose 4-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0