This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01834525_g1
See other REV1 GE Assays ›
Gene Symbol:
Rev1
Gene Name:
Rev1 DNA directed polymerase
Gene Aliases:
AAF47401, CG12189, DmREV1, DmRev1, Dmel\CG12189, Dmel_CG12189, REV1, dREV1, dRev1, drev1, rev1
Chromosome Location:
Chr.3L: 654363 - 657971 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0035150
Assay ID Dm01834525_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rev1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138203.3 NP_612047.1 6 - 7 2924 61
GenBank mRNA AB049435.1 - 6 - 7 2914 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
regulation of double-strand break repair via homologous recombination
translesion synthesis
error-prone translesion synthesis
DNA synthesis involved in double-strand break repair via homologous recombination
error-free translesion synthesis
damaged DNA binding
DNA-directed DNA polymerase activity
deoxycytidyl transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0