This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01836318_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
Rbfox1
Gene Name:
RNA-binding Fox protein 1
Gene Aliases:
A2BP1, A2bp1, BcDNA:GM06073, CG14155, CG18441, CG32062, Dmel\CG32062, Dmel_CG32062, cg18441, dA2BP1, rbfox1
Chromosome Location:
Chr.3L: 10481412 - 10594012 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0052062
Assay ID Dm01836318_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbfox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001104102.4 NP_001097572.1 5 - 6 1756 74
NM_001104103.3 NP_001097573.1 5 - 6 1741 74
NM_001104104.3 NP_001097574.2 6 - 7 1939 74
NM_001259778.1 NP_001246707.1 6 - 7 2462 74
NM_001259779.1 NP_001246708.1 5 - 6 2264 74
NM_001274751.1 NP_001261680.1 4 - 5 1264 74
NM_001370003.1 NP_001356980.1 5 - 6 1741 74
NM_168412.4 NP_729614.2 6 - 7 1939 74
NM_168413.3 NP_729615.2 6 - 7 1939 74
GenBank mRNA BT010058.1 - 5 - 6 1728 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
nervous system development
imaginal disc-derived wing vein specification
memory
negative regulation of translation
germarium-derived oocyte differentiation
positive regulation of transcription, DNA-templated
oogenesis
transcription regulatory region sequence-specific DNA binding
mRNA binding
mRNA 3'-UTR binding
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0