This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01836937_m1
See other CG43373 GE Assays ›
Gene Symbol:
CG43373
Gene Name:
CG43373-PB
Gene Aliases:
AC, Ago2, CG12436, CG32158, CG42513, Dmel\CG43373, Dmel_CG43373
Chromosome Location:
Chr.3L: 16491838 - 16543370 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263131
Assay ID Dm01836937_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CG43373
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001104152.4 NP_001097622.2 11 - 12 2494 75
NM_168670.4 NP_730173.3 11 - 12 2345 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

cAMP biosynthetic process
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
adenylate cyclase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline