This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01837530_g1
See other ATPSYNDELTA GE Assays ›
Gene Symbol:
ATPsyndelta
Gene Name:
ATP synthase, delta subunit
Gene Aliases:
ATP5D, ATPD, CG2968, Dmel\CG2968, Dmel_CG2968, anon-WO03040301.109, anon-WO03040301.110, l (1) G0230, l(1)G0230
Chromosome Location:
Chr.X: 10212970 - 10217393 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0028342
Assay ID Dm01837530_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATPsyndelta
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272468.1 NP_001259397.1 3 - 4 528 76
NM_001298145.1 NP_001285074.1 3 - 4 528 76
NM_132382.3 NP_572610.1 3 - 4 528 76
GenBank mRNA AY071203.1 - 3 - 4 526 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
hydrogen ion transmembrane transport
proton-transporting ATP synthase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline