Assay Details
| Gene Symbol: DNAlig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132679.3 | NP_572907.1 | 1 - 2 | 382 | 58 | |
| GenBank mRNA | BT057979.1 | - | 1 - 2 | 380 | 58 | |
More Information
Set Membership: |
|

DNAlig4
Genomic Map