This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01838631_g1
See other DNALIG4 GE Assays ›
Gene Symbol:
DNAlig4
Gene Name:
DNA ligase 4
Gene Aliases:
CG12176, Dmel\CG12176, Dmel_CG12176, Lig4, RE37186, lig4, ligIV, ligaseIV
Chromosome Location:
Chr.X: 13664584 - 13667811 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0030506
Assay ID Dm01838631_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNAlig4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_132679.3 NP_572907.1 2 - 3 552 69
GenBank mRNA BT057979.1 - 2 - 3 550 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
nucleotide-excision repair, DNA gap filling
double-strand break repair via nonhomologous end joining
DNA recombination
response to ionizing radiation
replication fork processing
DNA ligation involved in DNA repair
DNA biosynthetic process
DNA binding
DNA ligase (ATP) activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline