This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01840963_g1
See other CPSF6 GE Assays ›
Gene Symbol:
Cpsf6
Gene Name:
Cleavage and polyadenylation specific factor 6
Gene Aliases:
CF I[[m]]68, CFI68, CFIm68, CFlm-68k, CG7185, CPSF6, Dmel\CG7185, Dmel_CG7185, cg7185, dCPSF6
Chromosome Location:
Chr.3L: 8310493 - 8315502 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0035872
Assay ID Dm01840963_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259736.1 NP_001246665.1 3 - 4 1621 59
NM_139949.3 NP_648206.1 4 - 5 1762 59
GenBank mRNA AY058563.1 - 4 - 5 1740 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
positive regulation of autophagy
regulation of RNA splicing
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline