This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01841092_g1
See other GSTO1 GE Assays ›
Gene Symbol:
GstO1
Gene Name:
Glutathione S transferase O1
Gene Aliases:
CG6662, DmGSTO1, Dmel\CG6662, Dmel_CG6662, GSTO1, dGstO1
Chromosome Location:
Chr.3L: 8523742 - 8524866 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0035907
Assay ID Dm01841092_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GstO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139980.3 NP_648237.1 3 - 4 803 67
GenBank mRNA AY071499.1 - 3 - 4 798 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
oxidation-reduction process
glutathione transferase activity
pyrimidodiazepine synthase activity
transferase activity, transferring sulfur-containing groups
glutathione dehydrogenase (ascorbate) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0