This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842190_s1
See other RAC1 GE Assays ›
Gene Symbol:
Rac1
Gene Name:
CG2248-PA
Gene Aliases:
2248, CG2248, D-Rac, D-Rac 1, D-Rac1, DRac, DRac1, DRacA, Dm Rac1, DmRAC, DmRAC1, Dmel\CG2248, Dmel_CG2248, Dmrac1, Drac, Drac1, Drac1a, DracA, GTPase Rac1, P40792, RAC1, Rac, Rac GTPase, Rac-1, Rac3, RacA, dRac, dRac1, drac1, rac, rac-1, rac1
Chromosome Location:
Chr.3L: 1300879 - 1302683 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0010333
Assay ID Dm01842190_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001274318.1 NP_001261247.1 1 1548 147
NM_057602.4 NP_476950.1 1 1548 147
GenBank mRNA Z35642.1 - 1476 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
positive regulation of cell-matrix adhesion
immune response-regulating cell surface receptor signaling pathway involved in phagocytosis
actin filament organization
establishment or maintenance of cell polarity
JNK cascade
small GTPase mediated signal transduction
border follicle cell migration
germ-band shortening
dorsal closure
dorsal closure, elongation of leading edge cells
dorsal closure, spreading of leading edge cells
axon guidance
axonal fasciculation
ventral cord development
peripheral nervous system development
open tracheal system development
tracheal outgrowth, open tracheal system
salivary gland morphogenesis
hemocyte development
myoblast fusion
memory
motor neuron axon guidance
mesodermal cell migration
head involution
glial cell migration
regulation of cell shape
response to wounding
negative regulation of lamellipodium assembly
axon midline choice point recognition
muscle attachment
dendrite development
lamellipodium assembly
actin cytoskeleton organization
cortical cytoskeleton organization
cortical actin cytoskeleton organization
neuron projection development
actin cytoskeleton reorganization
activation of protein kinase activity
regulation of actin cytoskeleton organization
encapsulation of foreign target
melanotic encapsulation of foreign target
hemocyte migration
cell competition in a multicellular organism
imaginal disc-derived wing hair site selection
rhabdomere development
engulfment of apoptotic cell
positive regulation of axon extension
positive regulation of JNK cascade
dorsal closure, amnioserosa morphology change
dorsal appendage formation
vascular endothelial growth factor receptor signaling pathway
embryonic anterior midgut (ectodermal) morphogenesis
axon extension
muscle fiber development
neuron projection morphogenesis
regulation of dendrite morphogenesis
regulation of axonogenesis
regulation of synapse organization
sensory perception of touch
actin filament bundle assembly
myoblast proliferation
positive regulation of filopodium assembly
regulation of synapse assembly
positive regulation of protein serine/threonine kinase activity
determination of digestive tract left/right asymmetry
positive regulation of wound healing
nephrocyte filtration
positive regulation of axon guidance
regulation of adherens junction organization
positive regulation of border follicle cell migration
regulation of locomotor rhythm
GTPase activity
protein binding
GTP binding
protein kinase binding
GTPase activating protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0