This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842862_g1
See other ERF3 GE Assays ›
Gene Symbol:
eRF3
Gene Name:
eukaryotic translation release factor 3
Gene Aliases:
CG6382, DSUP35, Dmel\CG6382, Dmel_CG6382, Dsup35, Elf, deRF3, delf, dsup35, elf
Chromosome Location:
Chr.2L: 12434452 - 12441279 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0020443
Assay ID Dm01842862_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eRF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273486.1 NP_001260415.1 3 - 4 1932 68
NM_001273487.1 NP_001260416.1 3 - 4 1249 68
NM_001298945.1 NP_001285874.1 3 - 4 1492 68
NM_057911.4 NP_477259.1 3 - 4 1932 68
GenBank mRNA AY070979.1 - 3 - 4 1505 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational termination
translation
translational termination
translation release factor activity
GTPase activity
GTP binding
translation termination factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0