This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01843687_g1
See other SYX4 GE Assays ›
Gene Symbol:
Syx4
Gene Name:
Syntaxin 4
Gene Aliases:
CG2715, Dm Syx4, DmSyx4, Dmel\CG2715, Dmel_CG2715, EG:95B7.1, EG:95B7.11, STX4, Syx, Syx-4, syx4
Chromosome Location:
Chr.X: 2739089 - 2744076 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0024980
Assay ID Dm01843687_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Syx4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272266.1 NP_001259195.1 3 - 4 373 81
NM_080318.3 NP_525057.2 3 - 4 957 81
GenBank mRNA BT150201.1 - 3 - 4 949 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle fusion
regulation of synaptic growth at neuromuscular junction
synaptic vesicle docking
vesicle-mediated transport
vesicle docking
membrane fusion
retrograde trans-synaptic signaling
SNARE binding
SNAP receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline