Assay Details
| Gene Symbol: Gapdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272655.1 | NP_001259584.1 | 2 | 1252 | 144 | |
| NM_080714.4 | NP_542445.1 | 2 | 1318 | 144 | ||
| GenBank mRNA | AY094940.1 | - | 2 | 1318 | 144 | |
| Gene Symbol: CG1607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT029261.1 | - | 3229 | 144 | ||
More Information
Set Membership: |
|

Gapdh2
Genomic Map