This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02134761_m1
See other ACE GE Assays ›
Gene Symbol:
Ace
Gene Name:
Acetylcholine esterase
Gene Aliases:
ACE, AChE, AcChE, AchE, CG17907, CHE, DmAChE, Dm_ace, Dmel\CG17907, Dmel_CG17907, ace, ace-2, ache, dAChE, dmAChE, l(3)26, l(3)87Ed
Chromosome Location:
Chr.3R: 13222951 - 13259517 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000024
Assay ID Dm02134761_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ace
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170129.2 NP_001163600.1 8 - 9 2224 64
NM_001275601.1 NP_001262530.1 7 - 8 2511 64
NM_057605.5 NP_476953.1 7 - 8 2511 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

acetylcholine catabolic process in synaptic cleft
acetylcholine catabolic process
chemical synaptic transmission
choline catabolic process
acetylcholinesterase activity
cholinesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0